Learn More
Ace Glass, Inc. Flask, nephelo, 300mL, baffled, 14mm OD x 130mm sidearm, 25mm longneck, 210mL capacity below sidearm, cs/4
SDP
Catalog No. 50971454
Encompass_Preferred
Flask, nephelo, 300mL, baffled, 14mm OD x 130mm sidearm, 25mm longneck, 210mL capacity below sidearm, cs/4
Catalog No. 50-971-454 Supplier Ace Glass, Inc. Supplier No. 391205
Flask, nephelo, 300mL, baffled, 14mm OD x 130mm sidearm, 25mm longneck, 210mL capacity below sidearm, cs/4