Learn More
Ace Glass, Inc. Flask, nephelo, 300mL, single sidearm, 25mm OD long neck, 210mL capacity below sidearm, cs/4
SDP
Catalog No. 50971446
Encompass_Preferred
Flask, nephelo, 300mL, single sidearm, 25mm OD long neck, 210mL capacity below sidearm, cs/4
Catalog No. 50-971-446 Supplier Ace Glass, Inc. Supplier No. 390824
Flask, nephelo, 300mL, single sidearm, 25mm OD long neck, 210mL capacity below sidearm, cs/4