Learn More
Ace Glass, Inc. Flask, nephelo, 300mL, 12mm OD x 130mm sidearm, with screw caps cs/4
SDP
Catalog No. 50971447
Encompass_Preferred
300mL Nephelo culture flask with side port and angled sidearm cuvette for standard Nephelometers and spectrophotometers. Sidearm dimensions: 12mm O.D. x 130mm. Supplied with 38-460 phenolic top cap and 18-415 clean-out port cap.
Catalog No. 50-971-447 Supplier Ace Glass, Inc. Supplier No. 391003
300mL Nephelo culture flask with side port and angled sidearm cuvette for standard Nephelometers and spectrophotometers. Sidearm dimensions: 12mm O.D. x 130mm. Supplied with 38-460 phenolic top cap and 18-415 clean-out port cap.