Learn More
Chemglass Life Sciences Flask Only, 2, 000mL Single Neck Heavy Wall Wide Mouth Recovery Flask, GL80
SDP
Catalog No. 502733590
Encompass_Preferred
Heavy wall, wide mouth flask with a GL80 glass thread and a 24/40 connecting adapter. Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation. Unique design makes it easier to handle highly viscous liquids, powders, and solids. The large opening permits trouble-free emptying and cleaning and easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation. Complete code supplied complete with PTFE GL80 to 24/40 adapter (CG-1512-X-20). PTFE adapter has FDA grade silicone gasket behind a PTFE envelope type seal so that liquid or powders are only exposed to glass or PTFE.
Catalog No. 50-273-3590 Supplier Chemglass Life Sciences Supplier No. CG1512X12
Heavy wall, wide mouth flask with a GL80 glass thread and a 24/40 connecting adapter. Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation. Unique design makes it easier to handle highly viscous liquids, powders, and solids. The large opening permits trouble-free emptying and cleaning and easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation. Complete code supplied complete with PTFE GL80 to 24/40 adapter (CG-1512-X-20). PTFE adapter has FDA grade silicone gasket behind a PTFE envelope type seal so that liquid or powders are only exposed to glass or PTFE.