Learn More
Chemglass Life Sciences Spinner Flask, 100mL, Dimpled Bottom, Replacement Glass Flask ONLY
SDP
Catalog No. 501532123
Encompass_Preferred
Replacement vessel is supplied with black phenolic rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.
Catalog No. 50-153-2123 Supplier Chemglass Life Sciences Supplier No. CLS1401100
Replacement vessel is supplied with black phenolic rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.