Learn More
Chemglass Life Sciences Spinner Flask, 3000mL, Dimpled Bottom, Replacement Glass Flask ONLY
SDP
Catalog No. 501532126
Encompass_Preferred
Replacement vessel is supplied with black phenolic rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.
Catalog No. 50-153-2126 Supplier Chemglass Life Sciences Supplier No. CLS14013L
Replacement vessel is supplied with black phenolic rubber lined caps for the 18mm side arms, polypropylene caps and pouring rings for the GL-32 and GL-45 side arms.