Learn More
Thomas Scientific Laboratory Flask, PC, 1L, Flat Base, 50mm cap, Vented, Sterile
SDP
Catalog No. 501458472
Encompass
Foxx Life Sciences Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin that complies with stringent FDA and USP requirements. Our flasks can be used for cell cultures, microbial cultures, media preparation and storage. Flasks are available in 1liter and 2 liter sizes with flat bases for general laboratory use, and baffled bases for increased oxygenation add mixing. They have molded in graduation marks and additional 20% overhead space. All Flasks and Versa Caps™ are autoclavable at 15 PSI and 121°C for 20 minutes. Our flasks are compatible with all 50 mm Versa Caps™ and adapter inserts, enabling easy customization of top connections. Vented Versa Caps™ with 0.2 μm hydrophobic membrane are available separately as an accessory.
Catalog No. 50-145-8472 Supplier Thomas Scientific Supplier No. 1198J66
Foxx Life Sciences Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin that complies with stringent FDA and USP requirements. Our flasks can be used for cell cultures, microbial cultures, media preparation and storage. Flasks are available in 1liter and 2 liter sizes with flat bases for general laboratory use, and baffled bases for increased oxygenation add mixing. They have molded in graduation marks and additional 20% overhead space. All Flasks and Versa Caps™ are autoclavable at 15 PSI and 121°C for 20 minutes. Our flasks are compatible with all 50 mm Versa Caps™ and adapter inserts, enabling easy customization of top connections. Vented Versa Caps™ with 0.2 μm hydrophobic membrane are available separately as an accessory.