Learn More
Ace Glass, Inc. Flask, pear-shaped, 10mL, 14/20 polished joint
SDP
Catalog No. 50976920
Encompass_Preferred
10mL pear-shaped, single neck, heavy duty flask with a 14/20 polished joint.
Catalog No. 50-976-920 Supplier Ace Glass, Inc. Supplier No. 947747
10mL pear-shaped, single neck, heavy duty flask with a 14/20 polished joint.