Learn More
Ace Glass, Inc. Flask, pear-shaped, 50mL, 19/22 ground joint
SDP
Catalog No. 50976914
Encompass_Preferred
50mL pear-shaped, single neck, heavy duty flask with a 19/22 ground joint.
Catalog No. 50-976-914 Supplier Ace Glass, Inc. Supplier No. 947729
50mL pear-shaped, single neck, heavy duty flask with a 19/22 ground joint.