Learn More
Chemglass Life Sciences 10mL Single Neck Pear Shaped Flask, Minum-Ware
SDP
Catalog No. 50873222
Encompass_Preferred
10mL Single Neck Pear Shaped Flask, Minum-Ware. Pear-shaped single neck flask with 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).
Catalog No. 50-873-222 Supplier Chemglass Life Sciences Supplier No. MW5501
10mL Single Neck Pear Shaped Flask, Minum-Ware. Pear-shaped single neck flask with 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).