Learn More
Chemglass Life Sciences Flask, PFA, Round Bottom, 250mL, 29/32 Outer Joint Single Neck
SDP
Catalog No. 50872053
Encompass_Preferred
Flask, PFA, Round Bottom, 250mL, 29/32 Outer Joint Single Neck. All PFA single neck round bottom flask with standard taper outer joint. Flask can withstand MAX temperature of 250°C and a MIN temperature of -200°C.
Catalog No. 50-872-053 Supplier Chemglass Life Sciences Supplier No. CG1506T03
Flask, PFA, Round Bottom, 250mL, 29/32 Outer Joint Single Neck. All PFA single neck round bottom flask with standard taper outer joint. Flask can withstand MAX temperature of 250°C and a MIN temperature of -200°C.