Learn More
Chemglass Life Sciences Flask, PFA, Round Bottom, 250mL, 3-Neck, CN 29/32, SN 29/32
SDP
Catalog No. 501531148
Encompass_Preferred
Flask, PFA, Round Bottom, 250mL, 3-Neck, CN 29/32, SN 29/32. All PFA three neck round bottom flask with standard taper outer joint center neck and two additional 20 angled side necks. Allows assembly with a temperature probe, electrode or tube. Flask can withstand MAX temperature of 250C and a MIN temperature of -200C.
Catalog No. 50-153-1148 Supplier Chemglass Life Sciences Supplier No. CG1524T03
Flask, PFA, Round Bottom, 250mL, 3-Neck, CN 29/32, SN 29/32. All PFA three neck round bottom flask with standard taper outer joint center neck and two additional 20 angled side necks. Allows assembly with a temperature probe, electrode or tube. Flask can withstand MAX temperature of 250C and a MIN temperature of -200C.