Learn More
Chemglass Life Sciences Flask, Quartz, 1, 000mL, Flat Bottom, 29/42
SDP
Catalog No. 50872038
Encompass_Preferred
Flask, Quartz, 1,000mL, Flat Bottom, 29/42. Single neck, flat bottom, quartz flask with standard taper outer joint.
Catalog No. 50-872-038 Supplier Chemglass Life Sciences Supplier No. CG1500Q18
Flask, Quartz, 1,000mL, Flat Bottom, 29/42. Single neck, flat bottom, quartz flask with standard taper outer joint.