Learn More
DC Scientific Glass Inc DC Scientific Glass 125 mL Quartz Distillation Flask, ASTM D86 Application, Automated Analyzer Compatible
SDP
Catalog No. NC0754066
Encompass_Preferred
The DC Scientific Glass DCP-0086-006 is a 125 mL quartz distillation flask designed for ASTM D86 distillation analyses. Its high-quality quartz construction ensures superior thermal stability and chemical resistance, making it ideal for accurately distilling petroleum products. Compatible with automated D86 distillation analyzers, especially the OptiDist, this flask delivers reliable and reproducible results, differentiating it from other flasks in laboratory settings.
Catalog No. NC0754066 Supplier DC Scientific Glass Inc Supplier No. DCP0086006
The DC Scientific Glass DCP-0086-006 is a 125 mL quartz distillation flask designed for ASTM D86 distillation analyses. Its high-quality quartz construction ensures superior thermal stability and chemical resistance, making it ideal for accurately distilling petroleum products. Compatible with automated D86 distillation analyzers, especially the OptiDist, this flask delivers reliable and reproducible results, differentiating it from other flasks in laboratory settings.