Learn More
Chemglass Life Sciences Flask, Quartz, 150mL, Flat Bottom, 24/40
SDP
Catalog No. 50872033
Encompass_Preferred
Flask, Quartz, 150mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.
Catalog No. 50-872-033 Supplier Chemglass Life Sciences Supplier No. CG1500Q08
Flask, Quartz, 150mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.