Learn More
Chemglass Life Sciences Flask, Quartz, 250mL, Flat Bottom, 24/40
SDP
Catalog No. 50872035
Encompass_Preferred
Flask, Quartz, 250mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.
Catalog No. 50-872-035 Supplier Chemglass Life Sciences Supplier No. CG1500Q12
Flask, Quartz, 250mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.