Learn More
Chemglass Life Sciences Flask, Quartz, 500mL, Flat Bottom, 24/40
SDP
Catalog No. 501531118
Encompass_Preferred
Flask, Quartz, 500mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.
Catalog No. 50-153-1118 Supplier Chemglass Life Sciences Supplier No. CG1500Q14
Flask, Quartz, 500mL, Flat Bottom, 24/40. Single neck, flat bottom, quartz flask with standard taper outer joint.