Learn More
Chemglass Life Sciences Flask, Quartz, 500mL, Round Bottom, 24/40
SDP
Catalog No. 501531127
Encompass_Preferred
Flask, Quartz, 500mL, Round Bottom, 24/40. Quartz single neck round bottom flask with standard taper outer joint.
Catalog No. 50-153-1127 Supplier Chemglass Life Sciences Supplier No. CG1506Q14
Flask, Quartz, 500mL, Round Bottom, 24/40. Quartz single neck round bottom flask with standard taper outer joint.