Learn More
Wilmad Labglass FLASK RB 1N 300ML 24/40
Catalog No. 50803467
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 300mL capacity, with [ST] 24/40 outer joint.
Catalog No. 50-803-467 Supplier Wilmad Labglass Supplier No. LG7220150
Boiling Flask, Round Bottom, Short Neck, 300mL capacity, with [ST] 24/40 outer joint.