Learn More
Wilmad Labglass FLASK RB 1N 50ML 14/20
Catalog No. 50807906
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 50mL capacity, with [ST] 14/20 outer joint.
This item is not returnable. View return policy
Catalog No. 50-807-906 Supplier Wilmad Labglass Supplier No. ML1110718
Boiling Flask, Round Bottom, Short Neck, 50mL capacity, with [ST] 14/20 outer joint.