Learn More
Wilmad Labglass FLASK RB 1N 50ML 24/40
Catalog No. 50807854
Encompass_Preferred
Boiling Flask, Round Bottom, Short Neck, 50mL capacity, with [ST] 24/40 outer joint.
Catalog No. 50-807-854 Supplier Wilmad Labglass Supplier No. LG7220140
Boiling Flask, Round Bottom, Short Neck, 50mL capacity, with [ST] 24/40 outer joint.