Learn More
Wilmad Labglass FLASK RB 500ML 24/40C V24/40S
Catalog No. 50806544
Encompass_Preferred
Boiling Flask, Round Bottom, 2-Neck, 500mL capacity. Joint Sizes: Center Neck [ST] 24/40 outer; Vertical Side Neck: [ST] 24/40 outer. Round bottom two neck boiling flask with outer joints on both the center neck and the vertical side neck.
Catalog No. 50-806-544 Supplier Wilmad Labglass Supplier No. LG7290246
Boiling Flask, Round Bottom, 2-Neck, 500mL capacity. Joint Sizes: Center Neck [ST] 24/40 outer; Vertical Side Neck: [ST] 24/40 outer. Round bottom two neck boiling flask with outer joints on both the center neck and the vertical side neck.