Learn More
Chemglass Life Sciences Flask, Round Bottom, 5000mL, 3-Neck, Heavy Wall, 24/40 - 24/40, CG-911-A Drain Valve
SDP
Catalog No. 501311060
Encompass_Preferred
Flask, Round Bottom, 5000mL, 3-Neck, Heavy Wall, 24/40 - 24/40, CG-911-A Drain Valve. Heavy wall round bottom three neck flask with standard taper outer joints on center and side necks. Lower outlet is a CG-911-A extended tip which fills the seat area and permits total agitation of the contents of the flask. Once opened the contents of the flask will drain through the sidearm. The sidearm on the CG-911-A valve is 1/2" Swagelok.
Catalog No. 50-131-1060 Supplier Chemglass Life Sciences Supplier No. CG1523V05
Flask, Round Bottom, 5000mL, 3-Neck, Heavy Wall, 24/40 - 24/40, CG-911-A Drain Valve. Heavy wall round bottom three neck flask with standard taper outer joints on center and side necks. Lower outlet is a CG-911-A extended tip which fills the seat area and permits total agitation of the contents of the flask. Once opened the contents of the flask will drain through the sidearm. The sidearm on the CG-911-A valve is 1/2" Swagelok.