Learn More
Chemglass Life Sciences 1000mL Flask, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk
SDP
Catalog No. 50873072
Encompass_Preferred
1000mL Flask, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-873-072 Supplier Chemglass Life Sciences Supplier No. AF052826
1000mL Flask, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.