Learn More
Chemglass Life Sciences 500mL 2-Neck Flask, Reaction, 24/40 Joint Size, Valve, Airfree, Schlenk
SDP
Catalog No. 50873074
Encompass_Preferred
*AF053024 does not include stopcock. Please use AF053005 for flask with stopper* 500mL 2-Neck Flask, Reaction, 24/40 Joint Size, Valve, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.
Catalog No. 50-873-074 Supplier Chemglass Life Sciences Supplier No. AF053024
*AF053024 does not include stopcock. Please use AF053005 for flask with stopper* 500mL 2-Neck Flask, Reaction, 24/40 Joint Size, Valve, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.