Learn More
Chemglass Life Sciences 50mL Flask, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF052820
Encompass_Preferred
50mL Flask, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-153-0969