Learn More
Chemglass Life Sciences 100mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask
SDP
Catalog No. CG151257
Encompass_Preferred
100mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. CG151257 Supplier Chemglass Life Sciences Supplier No. CG151257
100mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.