Learn More
Chemglass Life Sciences 25mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask
SDP
Catalog No. CG151253
Encompass_Preferred
25mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
This item is not returnable. View return policy
Catalog No. CG151253 Supplier Chemglass Life Sciences Supplier No. CG151253
25mL Single Neck Evaporating Flask, 14/20 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.