Learn More
Chemglass Life Sciences 200mL Single Neck Evaporating Flask, 19/22 Outer Joint, Recovery Flask
SDP
Catalog No. CG151260
Encompass_Preferred
200mL Single Neck Evaporating Flask, 19/22 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. CG151260 Supplier Chemglass Life Sciences Supplier No. CG151260
200mL Single Neck Evaporating Flask, 19/22 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.