Learn More
Ace Glass, Inc. Flask, receiving, 10L, all R220 models, part 46519
SDP
Catalog No. 50978874
Encompass
10L borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 46519.
Catalog No. 50-978-874 Supplier Ace Glass, Inc. Supplier No. 672615
10L borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 46519.