Learn More
Ace Glass, Inc. Flask, receiving, 10L, ambered, all R220 models
SDP
Catalog No. 50973406
Encompass
10L ambered borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring.
Catalog No. 50-973-406 Supplier Ace Glass, Inc. Supplier No. 672630
10L ambered borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring.