Learn More
Ace Glass, Inc. Flask, receiving, 10L, poly-coated, all R220 models, part 37569
SDP
Catalog No. 50973403
Encompass
10L poly-coated borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 37569.
Catalog No. 50-973-403 Supplier Ace Glass, Inc. Supplier No. 672610
10L poly-coated borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 37569.