Learn More
Ace Glass, Inc. Flask, receiving, 20L, all R220 models, part 28671
SDP
Catalog No. 50973405
Encompass
20L borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 28671.
Catalog No. 50-973-405 Supplier Ace Glass, Inc. Supplier No. 672625
20L borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 28671.