Learn More
Ace Glass, Inc. Flask, receiving, 20L, ambered, all R220 models
SDP
Catalog No. 50978875
Encompass
20L ambered borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring.
Catalog No. 50-978-875 Supplier Ace Glass, Inc. Supplier No. 672632
20L ambered borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring.