Learn More
Ace Glass, Inc. Flask, receiving, 20L, poly-coated, all R220 models, part 41446
SDP
Catalog No. 50973404
Encompass
20L poly-coated borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 41446.
Catalog No. 50-973-404 Supplier Ace Glass, Inc. Supplier No. 672620
20L poly-coated borosilicate receiving flask for all R220 model rotary evaporators. Side neck includes an SVL thread vent cap. Bottom inner ball joint includes a CAPFE o-ring. Replaces Part number 41446.