Learn More
Wilmad Labglass FLASK RECOVERY 1000ML 24/40
Catalog No. 50803371
Encompass_Preferred
Recovery Flask, 1000mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-803-371 Supplier Wilmad Labglass Supplier No. LG7245108
Recovery Flask, 1000mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.