Learn More
Wilmad Labglass FLASK RECOVERY 100ML 24/40
Catalog No. 50807383
Encompass_Preferred
Recovery Flask, 100mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-807-383 Supplier Wilmad Labglass Supplier No. LG7245102
Recovery Flask, 100mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.