Learn More
Wilmad Labglass FLASK RECOVERY 10ML 14/20
Catalog No. 50807636
Encompass_Preferred
Boiling Flask, Recovery, 10mL Capacity with [ST] 14/20 Outer Joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-807-636 Supplier Wilmad Labglass Supplier No. ML1155700
Boiling Flask, Recovery, 10mL Capacity with [ST] 14/20 Outer Joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.