Learn More
Ace Glass, Inc. Flask, recovery, 1L, 24/40, indented, Buchi; 00424
SDP
Catalog No. 50971537
Encompass_Preferred
1L replacement recovery flask for Buchi 00424 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.
Catalog No. 50-971-537 Supplier Ace Glass, Inc. Supplier No. 399422
1L replacement recovery flask for Buchi 00424 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.