Learn More
Ace Glass, Inc. Flask, recovery, 1L, 29/32, indented, Buchi; 00453
SDP
Catalog No. 50978251
Encompass_Preferred
1L replacement recovery flask for Buchi 00453 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.
Catalog No. 50-978-251 Supplier Ace Glass, Inc. Supplier No. 399412
1L replacement recovery flask for Buchi 00453 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.