Learn More
Wilmad Labglass FLASK RECOVERY 200ML 24/40
Catalog No. 50803919
Encompass_Preferred
Recovery Flask, 200mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.
Catalog No. 50-803-919 Supplier Wilmad Labglass Supplier No. LG7245104
Recovery Flask, 200mL capacity, with [ST] 24/40 outer joint. Upper portion of the flask has a gradual taper making it easier to remove reaction products. Lower hemisphere fits comparably sized heating mantles.