Learn More
Chemglass Life Sciences 25mL Recovery Flask, Minum-Ware
SDP
Catalog No. 50873227
Encompass_Preferred
25mL Recovery Flask, Minum-Ware. Kjeldahl shaped single neck flask with a 14/10 threaded outer joint. Sloped sides permit easy removal of product with a spatula. Supplied complete with an open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).
Catalog No. 50-873-227 Supplier Chemglass Life Sciences Supplier No. MW5702
25mL Recovery Flask, Minum-Ware. Kjeldahl shaped single neck flask with a 14/10 threaded outer joint. Sloped sides permit easy removal of product with a spatula. Supplied complete with an open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).