Learn More
Ace Glass, Inc. Flask, recovery, 2L, 24/40, indented, Buchi; 11580
SDP
Catalog No. 50978252
Encompass_Preferred
2L replacement recovery flask for Buchi 11580 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.
Catalog No. 50-978-252 Supplier Ace Glass, Inc. Supplier No. 399423
2L replacement recovery flask for Buchi 11580 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.