Learn More
Ace Glass, Inc. Flask, recovery, 2L, 24/40, indented, safety coated, Buchi;
SDP
Catalog No. 50971542
Encompass_Preferred
2L safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.
Catalog No. 50-971-542 Supplier Ace Glass, Inc. Supplier No. 3994126
2L safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.