Learn More
Ace Glass, Inc. Flask, recovery, 2L, 29/32, indented, Buchi; 00454
SDP
Catalog No. 50971535
Encompass_Preferred
2L replacement recovery flask for Buchi 00454 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.
Catalog No. 50-971-535 Supplier Ace Glass, Inc. Supplier No. 399414
2L replacement recovery flask for Buchi 00454 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.