Learn More
Chemglass Life Sciences 3000ml Single Neck Evaporating Flask, 29/42 outer joint, Recovery Flask
SDP
Catalog No. 50873105
Encompass_Preferred
3000ml Single Neck Evaporating Flask, 29/42 outer joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. 50-873-105 Supplier Chemglass Life Sciences Supplier No. CG151246
3000ml Single Neck Evaporating Flask, 29/42 outer joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.