Learn More
Ace Glass, Inc. Flask, recovery, 500mL, 24/40, indented, Buchi; 11579
SDP
Catalog No. 50971536
Encompass_Preferred
500mL replacement recovery flask for Buchi 11579 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.
Catalog No. 50-971-536 Supplier Ace Glass, Inc. Supplier No. 399420
500mL replacement recovery flask for Buchi 11579 rotary evaporator. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.