Learn More
Ace Glass, Inc. Flask, recovery, 500mL, 24/40, indented, safety coated, Buchi;
SDP
Catalog No. 50978253
Encompass_Preferred
500mL safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.
Catalog No. 50-978-253 Supplier Ace Glass, Inc. Supplier No. 3994120
500mL safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 24/40 standard taper single neck.