Learn More
Ace Glass, Inc. Flask, recovery, 500mL, 29/32, indented, safety coated, Buchi;
SDP
Catalog No. 50971538
Encompass_Preferred
500mL safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.
Catalog No. 50-971-538 Supplier Ace Glass, Inc. Supplier No. 3994110
500mL safety-coated replacement recovery flask for Buchi rotary evaporators. Flask is indented, pear shaped with a heavy wall, and features a 29/32 standard taper single neck.