Learn More
Ace Glass, Inc. FLASK,RECOVERY,HEAVY WALL,3L
SDP
Catalog No. NC2231367
Encompass_Preferred
Flask,recovery,heavy wall,3L,24/40A multipurpose, round bottom flask with a single neck and sloping side wall. Designed for use as a recovery flask with rotary evaporators. Heavy wall product family members feature approximately 30% heavier wall weights.
This item is not returnable. View return policy
Catalog No. NC2231367 Supplier Ace Glass, Inc. Supplier No. 6892249
Flask,recovery,heavy wall,3L,24/40A multipurpose, round bottom flask with a single neck and sloping side wall. Designed for use as a recovery flask with rotary evaporators. Heavy wall product family members feature approximately 30% heavier wall weights.